Learn More
GE Healthcare illustra™ TempliPhi™ 100/500 Amplification Kit
Special Offer! (Expires 12/31/2015)
Manufacturer:  GE Healthcare 29168043
 View more versions of this product
Catalog No. 29-168-043
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Recommended Storage
Store at -20°C for one month or at -70°C until the expiration date on package.
Description & Specifications
Specifications
Prepare circular DNA templates for cycle sequencing, cloning and transformation
0.5mL Sample Buffer, 0.5mL Reaction Buffer, 20µL Enzyme mix and 50µL positive control pUC19 DNA
500 Reactions