GE Healthcare illustra™ TempliPhi™ 100/500 Amplification Kit

Special Offer! (Expires 12/31/2015)

Manufacturer: GE Healthcare 29168042

 View more versions of this product

Catalog No. 29-168-042

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    For Use With (Application) Prepare circular DNA templates for cycle sequencing, cloning and transformation
    Includes 0.5mL Sample Buffer, 0.5mL Reaction Buffer, 20µL Enzyme mix and 50µL positive control pUC19 DNA
    Quantity 100 Reactions

    Recommended Storage

    Store at -20°C for one month or at -70°C until the expiration date on package.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit