Learn More
GE Healthcare illustra™ triplePrep Kit : PROMOTION
Promotion Details: Receive a 28% discount.
Isolate gDNA, total RNA, and total denatured proteins from undivided animal tissues and mammalian cell samples in less than 1 hour
Manufacturer:  GE Healthcare 29361647
Catalog No. 45-001-810PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • High-yield process allows extraction/isolation from single limited-size samples from biopsies, archived tissues, and tumors
  • Isolated products suitable for downstream applications such as PCR, restriction digestion, sequencing, array CGH, RT-qPCR, gene expression microarray, SDS-PAGE, Western blotting, 2-D DIGE, and LCMS
  • Flexible workflow allows easy isolation of any two or all three analytes
  • Color-coded caps and bottles match to protocol steps to minimize the chance for error
Specifications
Specifications
Extraction and isolation of high quality genomic DNA, total RNA and total denatured Proteins from animal tissues and mammalian cells
Protocol and a supply of components sufficient for 50 preps
Kit
50 Preps