Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ILT1 (CD85h) Mouse anti-Human, PE, Clone: 24/ILT1 (also known as 24), BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566577
Description
ILT1 (CD85h) Monoclonal antibody specifically detects ILT1 (CD85h) in Human samples. It is validated for Flow Cytometry.Specifications
ILT1 (CD85h) | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Human | |
Purified |
Flow Cytometry | |
24/ILT1 (also known as 24) | |
PE | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2b κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title