Learn More
ILT2/CD85j/LILRB1 Mouse anti-Human, Janelia Fluor 549, Clone: HP-F1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142998JF549
Catalog No. NBP14298549
Add to cart
Description
Description
ILT2/CD85j/LILRB1 Monoclonal antibody specifically detects ILT2/CD85j/LILRB1 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
ILT2/CD85j/LILRB1
HP-F1
Flow Cytometry
Purified
LILRB1
The immunogen for this antibody was CD85j.
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CD85 antigen-like family member J, CD85j, CD85j antigen, Ig-like transcript 2, ILT-2, ILT2FLJ37515, Immunoglobulin-like transcript 2, leukocyte Ig-like receptor-1, Leukocyte immunoglobulin-like receptor 1, leukocyte immunoglobulin-like receptor subfamily B member 1, leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR1LIR-1MIR-7CD85MIR7CD85 antigen, member 1, Monocyte/macrophage immunoglobulin-like receptor 7
Mouse
IgG1 Kappa
0.1mL
Primary
10859