Learn More
Novus Biologicals™ ILT5/CD85a/LILRB3 Antibody Pair [HRP]
Polyclonal/Monoclonal Antibody Pair
Manufacturer:  Novus Biologicals™ NBP2793805PLATES
 View more versions of this product
Catalog No. NB009800
Add to cart
Description
Description
This solid phase sandwich ELISA utilizes an antibody specific for Human ILT5/CD85a/LILRB3.
Specifications
Specifications
Sandwich ELISA
Antibody Pairs
78.125 pg/mL
11025
RUO
ILT5/CD85a/LILRB3
5 Plates
78.125-5000 pg/mL
CD85 antigen-like family member A, CD85a antigen, HL9Immunoglobulin-like transcript 5, ILT-5, ILT5CD85a, Leukocyte immunoglobulin-like receptor 3, leukocyte immunoglobulin-like receptor subfamily B member 3, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR3CD85A, LIR-3MGC138403, member 3, Monocyte inhibitory receptor HL9, PIRB
HRP
This solid phase sandwich ELISA utilizes an antibody specific for Human ILT5/CD85a/LILRB3.
Sandwich ELISA
  • Standard: Each vial contains 200 ng of recombinant Human ILT5/CD85a/LILRB3.
  • Capture Antibody: 0.15 mg/mL of mouse anti-Human ILT5/CD85a/LILRB3 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human ILT5/CD85a/LILRB3 polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Storage is content dependent.
    Rabbit/Mouse
    Human
    LILRB3
    1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.8 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 5000 pg/mL is recommended.