Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ILT5/CD85a/LILRB3 Mouse anti-Human, Alexa Fluor™ 532, Clone: MM0413-9S32, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211729AF532
Description
ILT5/CD85a/LILRB3 Monoclonal specifically detects ILT5/CD85a/LILRB3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ILT5/CD85a/LILRB3 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
LILRB3 | |
Human recombinant ILT5/CD85a/LILRB3 | |
Primary | |
Human | |
IgG2 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0413-9S32 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CD85 antigen-like family member A, CD85a antigen, HL9Immunoglobulin-like transcript 5, ILT-5, ILT5CD85a, Leukocyte immunoglobulin-like receptor 3, leukocyte immunoglobulin-like receptor subfamily B member 3, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR3CD85A, LIR-3MGC138403, member 3, Monocyte inhibitory receptor HL9, PIRB | |
Mouse | |
0.1 mL | |
11025 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title