Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILT5/CD85a/LILRB3 Antibody (MM0413-9S32) - Azide and BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$204.00 - $542.50

Specifications

Antigen ILT5/CD85a/LILRB3
Clone MM0413-9S32
Concentration LYOPH
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018868
SDP
View Documents
Novus Biologicals
NBP2117290.025MG
0.025 mg
Each for $204.00
Add to Cart
 
NBP211729
SDP
View Documents
Novus Biologicals
NBP211729
0.1 mg
Each for $542.50
Add to Cart
 
Description

Description

ILT5/CD85a/LILRB3 Monoclonal specifically detects ILT5/CD85a/LILRB3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications

Specifications

ILT5/CD85a/LILRB3
LYOPH
Unconjugated
Mouse
Human
CD85 antigen-like family member A, CD85a antigen, HL9Immunoglobulin-like transcript 5, ILT-5, ILT5CD85a, Leukocyte immunoglobulin-like receptor 3, leukocyte immunoglobulin-like receptor subfamily B member 3, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR3CD85A, LIR-3MGC138403, member 3, Monocyte inhibitory receptor HL9, PIRB
LILRB3
Protein G purified
MM0413-9S32
Monoclonal
Purified
RUO
AAB68668
11025
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
SDS
Documents

Documents

Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit