Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ ILT6/CD85e/LILRA3 Antibody Pair [HRP]
Polyclonal/Monoclonal Antibody Pair
$1183.00
Specifications
Assay Range | 156.25 pg/mL |
---|---|
Assay Sensitivity | 156.25-10000 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Product Type | Antibody Pairs |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB009871
|
Novus Biologicals™
NBP27941615PLATES |
15 Plates |
Each of 1 for $1,183.00
|
|
|||||
Description
This solid phase sandwich ELISA utilizes an antibody specific for Human ILT6/CD85e/LILRA3.Specifications
156.25 pg/mL | |
Sandwich ELISA | |
Antibody Pairs | |
Human | |
CD85 antigen-like family member E, CD85e antigen, HM31, ILT-6, ILT6CD85e, Immunoglobulin-like transcript 6, leucocyte immunoglobulin-like receptor, Leukocyte immunoglobulin-like receptor 4, leukocyte immunoglobulin-like receptor A3, leukocyte immunoglobulin-like receptor subfamily A member 3, leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member3, LIR4CD85E, LIR-4HM43e3, Monocyte inhibitory receptor HM43/HM31 | |
LILRA3 | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 10000 pg/mL is recommended. |
156.25-10000 pg/mL | |
HRP | |
This solid phase sandwich ELISA utilizes an antibody specific for Human ILT6/CD85e/LILRA3. | |
Sandwich ELISA | |
11026 | |
Rabbit/Mouse | |
RUO | |
ILT6/CD85e/LILRA3 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title