Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ILT7/CD85g/LILRA4 Mouse anti-Human, Alexa Fluor 350, Clone: 656688, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB6287U100UG
This item is not returnable.
View return policy
Description
ILT7/CD85g/LILRA4 Monoclonal specifically detects ILT7/CD85g/LILRA4 in Human samples. It is validated for Flow Cytometry.Specifications
ILT7/CD85g/LILRA4 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P59901 | |
LILRA4 | |
Mouse myeloma cell line NS0-derived recombinant human LILRA4/CD85g/ILT7 Glu24-Asn446 Accession # P59901 | |
RUO | |
23547 | |
Human | |
IgG2a |
Flow Cytometry | |
656688 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD85 antigen-like family member G, CD85g, CD85g antigen, ILT7, ILT-7, ILT7MGC129598, Immunoglobulin-like transcript 7, leukocyte immunoglobulin-like receptor subfamily A member 4, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 4, leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member4, MGC129597 | |
Mouse | |
100 μg | |
Primary | |
Detects human LILRA4/CD85g/ILT7 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) LILRA5, rhILT2, 3, 4, 5, or 6 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)