Learn More
ILT8/CD85b/LILRA6 Mouse anti-Human, Alexa Fluor 405, Clone: 921330, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8656V100UG
Catalog No. FAB8656V100
Add to cart
This item is not returnable. View return policy
Description
Description
ILT8/CD85b/LILRA6 Monoclonal antibody specifically detects ILT8/CD85b/LILRA6 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
ILT8/CD85b/LILRA6
Monoclonal
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Mouse
IgG1
RUO
Primary
79168
Human
Flow Cytometry
921330
Flow Cytometry 0.25-1 ug/10^6 cells
Q6PI73
HEK293 human embryonic kidney cell line-derived recombinant human LILRA6/CD85b Met1-Asn447 Accession # Q6PI73
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human LILRA6/CD85b in direct ELISA. Detects HEK293 cells transfected with LILRA6/CD85b but not irrelevant transfectants.
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)