Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILVBL Mouse anti-Human, Rat, Canine, Monkey, Allophycocyanin/Cy7, Clone: OTI8B12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271661APCCY7

Catalog No. N27161APCY7


Add to cart

Description

Description

ILVBL Monoclonal antibody specifically detects ILVBL in Human, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

ILVBL
OTI8B12
Flow Cytometry
Affinity Purified, Conjugated
ILVBL
Full length recombinant protein of human ILVBL (NP_006835) produced in HEK293T cells.
Immunogen affinity purified
Epigenetics
Primary
10994.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
acetolactate synthase homolog, acetolactate synthase-like protein, AHASFLJ39061,209L8, EC 2.2.1, EC 2.2.1.-, ILV2H, ilvB (bacterial acetolactate synthase)-like, IlvB-like protein, MGC1269, MGC19535
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human, Monkey, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit