Learn More
Invitrogen™ Image-iT™ Fixative Solution (4% formaldehyde, methanol-free)
Invitrogen™ Image-iT™ Fixative Solution is a ready-to-use neutral PBS solution containing 4% formaldehyde (w/v) that is free of methanol.
$54.25 - $183.00
Specifications
Product Line Image-iT™
Reagent Type Fix Reagent
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FB002 Non-distribution product as customer accommodation.
View Documents
Invitrogen™
FB002
20 mL
Each for $54.25
Add to cart
R37814 Non-distribution product as customer accommodation.
View Documents
Invitrogen™
R37814
5 x 20 mL
Each for $183.00
Add to cart
Description
Description
  • Ready-to-use, with room temperature storage
  • High-purity 4% formaldehyde in PBS at neutral pH
  • Methanol-free
  • Optimized for secondary antibody labeling, fixed-cell dye staining, and GFP applications
Specifications
Specifications
Image-iT™
Fix Reagent