Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IMD 0354, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Inhibitor of IκB kinase-β (IKKβ) that blocks NF-κB nuclear translocation. Attenuates myocardial ischemia/reperfusion injury by decreasing expression of adhesion molecules ICAM-1 and P-selectin and inhibiting cytokine and chemokine production in cardiomyocytes. Induces G0/G1 cell cycle arrest and apoptosis in HMC-1 and breast cancer cells. Also antagonist at P2X1, P2X4 and P2X7 receptors (IC50 values are 19, 156 and 175 nM respectively). Inhibits platelet aggregation induced by collegen in vitro.
Specifications
Quantity | 10 mg |
Formula Weight | 383.67 |
Percent Purity | >99% |
Chemical Name or Material | IMD 0354 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction