Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imidazo[1,2-a]pyridine-6-carboxylic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
139022-25-6
C8H6N2O2
162.15
ONOJJCTXSDBVSP-UHFFFAOYSA-N
imidazo 1,2-a pyridine-6-carboxylic acid,h-imidazo 1,2-a pyridine-6-carboxylic acid,imidazo 1,2-a pyridine-6-carboxylicacid,1h-imidazo 1,2-a pyridine-6-carboxylic acid,4-hydroimidazo 1,2-a pyridine-6-carboxylic acid,pubchem23163,acmc-209cig,ksc173s9l,6-carboxyimidazo 1,2-a pyridine,imidazol 1,2-a pyridine-6-carboxylic acid
7075376
imidazo[1,2-a]pyridine-6-carboxylic acid
C1=CC2=NC=CN2C=C1C(=O)O
139022-25-6
C8H6N2O2
162.15
ONOJJCTXSDBVSP-UHFFFAOYSA-N
imidazo 1,2-a pyridine-6-carboxylic acid,h-imidazo 1,2-a pyridine-6-carboxylic acid,imidazo 1,2-a pyridine-6-carboxylicacid,1h-imidazo 1,2-a pyridine-6-carboxylic acid,4-hydroimidazo 1,2-a pyridine-6-carboxylic acid,pubchem23163,acmc-209cig,ksc173s9l,6-carboxyimidazo 1,2-a pyridine,imidazol 1,2-a pyridine-6-carboxylic acid
7075376
imidazo[1,2-a]pyridine-6-carboxylic acid
C1=CC2=NC=CN2C=C1C(=O)O
Melting Point 250°C
Quantity 10g
Packaging Glass bottle
Assay Percent Range 96% min. (HPLC)
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material Imidazo[1, 2-a]pyridine-6-carboxylic acid

GHS Signal Word: Warning