Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imidazo[1,2-a]pyridine-7-carbohydrazide, 95%, Thermo Scientific Chemicals

Catalog No. AAH35365MD
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 421595-78-0
Molecular Formula C8H8N4O
Molecular Weight (g/mol) 176.179
MDL Number MFCD13183226
InChI Key VXJBLZXLLRYHPD-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine-7-carbohydrazide, imidazo 1,2-a pyridine-7-carboxylic acid hydrazide, 7-hydrazinocarbonyl imidazo 1,2-a pyridine
PubChem CID 55281632
IUPAC Name imidazo[1,2-a]pyridine-7-carbohydrazide
SMILES C1=CN2C=CN=C2C=C1C(=O)NN

Specifications

Melting Point 258°C to 261°C
Quantity 250 mg
Formula Weight 176.18
Percent Purity 95%
Chemical Name or Material Imidazo[1,2-a]pyridine-7-carbohydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.