Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Imidazole, 0.2M buffer soln., pH 6.0, low endotoxin

Click to view available options
Quantity:
100mL
250mL
288-32-4
C3H4N2
68.079
MFCD00005183
RAXXELZNTBOGNW-UHFFFAOYSA-N
795
CHEBI:16069
1H-imidazole
C1=CN=CN1
288-32-4
C3H4N2
68.079
MFCD00005183
RAXXELZNTBOGNW-UHFFFAOYSA-N
795
CHEBI:16069
1H-imidazole
C1=CN=CN1
Concentration 0.2M buffer soln.
Physical Form Liquid
pH 6.0
Color Colorless
Quantity 100mL
Chemical Name or Material Imidazole, low endotoxin

TSCA : TSCA

storageNote1 : 2° to 8°C