Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Imidazole-4-acetic acid monohydrochloride, 97%

Click to view available options
Quantity:
1g
5g
3251-69-2
C5H7ClN2O2
162.573
MFCD00012698
MWHLCFYPFGFBQO-UHFFFAOYSA-N
4-imidazoleacetic acid hydrochloride,2-1h-imidazol-5-yl acetic acid hydrochloride,imidazole-4-acetic acid hydrochloride,2-1h-imidazol-4-yl acetic acid hydrochloride,4-imidazoleacetic acid hcl,imidazoleacetic acid hydrochloride,unii-et7a74roao,4-imidazoleaceticacidhydrochloride,i4aa,et7a74roao
145685
2-(1H-imidazol-5-yl)acetic acid;hydrochloride
C1=C(NC=N1)CC(=O)O.Cl
3251-69-2
C5H7ClN2O2
162.573
MFCD00012698
MWHLCFYPFGFBQO-UHFFFAOYSA-N
4-imidazoleacetic acid hydrochloride,2-1h-imidazol-5-yl acetic acid hydrochloride,imidazole-4-acetic acid hydrochloride,2-1h-imidazol-4-yl acetic acid hydrochloride,4-imidazoleacetic acid hcl,imidazoleacetic acid hydrochloride,unii-et7a74roao,4-imidazoleaceticacidhydrochloride,i4aa,et7a74roao
145685
2-(1H-imidazol-5-yl)acetic acid;hydrochloride
C1=C(NC=N1)CC(=O)O.Cl
Applications
A GABAc antagonistImidazole-4-acetic acid monohydrochloride is used in the preparation of imidazolyl-polyethylenimine modified nanoparticles. It is also used to prepare pyridyl and imidazoyl functionalized carboproteins and potential metal ion chelators. Further, it acts as an imidazole derivative with potential antituberculousis properties. In addition to this, it is used in the generation of acyl guanidine inhibitors of beta-secretas.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool place. Incompatible with oxidizing agents.
Quantity 1g
Odor Odorless
Melting Point 221°C to 224°C
Beilstein 3701591
Formula Weight 162.58
Percent Purity 97%
Chemical Name or Material Imidazole-4-acetic acid monohydrochloride