Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imidazole, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
100g
1kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
288-32-4
C3H4N2
68.079
RAXXELZNTBOGNW-UHFFFAOYSA-N
795
CHEBI:16069
1H-imidazole
C1=CN=CN1
288-32-4
C3H4N2
68.079
RAXXELZNTBOGNW-UHFFFAOYSA-N
795
CHEBI:16069
1H-imidazole
C1=CN=CN1

High quality chemicals for laboratory and specialized industrial use

Specific Gravity 1L = 0.60kg
Packaging Wide Mouth Amber Glass Bottle
Formula Weight 68.08
Flash Point 146.00°C
Quantity 100g
Grade Baker Analyzed™ ACS Reagent
Chemical Name or Material Imidazole