Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Imidazole, Molecular Biology Grade, ≥99%

Click to view available options
Quantity:
50g
100g
288-32-4
C3H4N2
68.079
MFCD00005183
RAXXELZNTBOGNW-UHFFFAOYSA-N
imidazole,glyoxaline,imidazol,iminazole,miazole,1,3-diazole,glyoxalin,imutex,1,3-diaza-2,4-cyclopentadiene,pyrro b monazole
795
CHEBI:16069
1H-imidazole
C1=CN=CN1
288-32-4
C3H4N2
68.079
MFCD00005183
RAXXELZNTBOGNW-UHFFFAOYSA-N
imidazole,glyoxaline,imidazol,iminazole,miazole,1,3-diazole,glyoxalin,imutex,1,3-diaza-2,4-cyclopentadiene,pyrro b monazole
795
CHEBI:16069
1H-imidazole
C1=CN=CN1
Melting Point 90°C
Density 1.03g/mL
Quantity 50g
Boiling Point 256°C
Color White
UN Number UN2923
Beilstein 103853
Merck Index 144,912
Formula Weight 68.08
Physical Form Powder
Percent Purity ≥99%
Grade Molecular Biology
Chemical Name or Material Imidazole
Show More Show Less

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

RTECSNumber : NI3325000

TSCA : TSCA