Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iminodibenzyl 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
494-19-9
C14H13N
195.26
MFCD00005070
ZSMRRZONCYIFNB-UHFFFAOYSA-N
iminodibenzyl,10,11-dihydro-5h-dibenzo b,f azepine,10,11-dihydro-5h-dibenz b,f azepine,iminobibenzyl,2,2'-iminodibenzyl,2,2'-iminobibenzyl,iminodibenyl,5h-dibenz b,f azepine, 10,11-dihydro,unii-262bx7oe3u,10,11-dihydro-5-dibenz b,f azepine
10308
6,11-dihydro-5H-benzo[b][1]benzazepine
C1CC2=CC=CC=C2NC3=CC=CC=C31
494-19-9
C14H13N
195.26
MFCD00005070
ZSMRRZONCYIFNB-UHFFFAOYSA-N
iminodibenzyl,10,11-dihydro-5h-dibenzo b,f azepine,10,11-dihydro-5h-dibenz b,f azepine,iminobibenzyl,2,2'-iminodibenzyl,2,2'-iminobibenzyl,iminodibenyl,5h-dibenz b,f azepine, 10,11-dihydro,unii-262bx7oe3u,10,11-dihydro-5-dibenz b,f azepine
10308
6,11-dihydro-5H-benzo[b][1]benzazepine
C1CC2=CC=CC=C2NC3=CC=CC=C31
Sulfated Ash 0.3% max.
Loss on Drying 0.5% max.
Packaging Glass bottle
Color Beige to Yellow
Infrared Spectrum Authentic
Melting Point 105°C to 108°C
Quantity 25g
Assay Percent Range 97%
Formula Weight 195.26
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material Iminodibenzyl, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 207-787-1

RTECSNumber : HN8950000

TSCA : TSCA