Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imipenem monohydrate, 95%, Alfa Aesar™

Click to view available options
Quantity:
5mg
74431-23-5
C12H19N3O5S
317.36
MFCD09753321
GSOSVVULSKVSLQ-JJVRHELESA-N
Primaxin; Tienam
5282372
CHEBI:51799
(5R,6S)-3-[2-(aminomethylideneamino)ethylsulfanyl]-6-[(1R)-1-hydroxyethyl]-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid;hydrate
CC(C1C2CC(=C(N2C1=O)C(=O)O)SCCN=CN)O.O
74431-23-5
C12H19N3O5S
317.36
MFCD09753321
GSOSVVULSKVSLQ-JJVRHELESA-N
Primaxin; Tienam
5282372
CHEBI:51799
(5R,6S)-3-[2-(aminomethylideneamino)ethylsulfanyl]-6-[(1R)-1-hydroxyethyl]-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid;hydrate
CC(C1C2CC(=C(N2C1=O)C(=O)O)SCCN=CN)O.O

Broad spectrum B-lactam antibiotic. It is a member of the carbapenem class of “magic bullet” antibiotics for severe infections.

Applications
Imipenem monohydrate is a broad spectrum B-lactam antibiotic. It is a member of the carbapenem class of “magic bullet“ antibiotics for severe infections.

Solubility
Soluble in water at 5mg/ml

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Merck Index 14,4919
Solubility Information Soluble in water at 5mg/mL
Formula Weight 317.36
Physical Form Powder
Percent Purity 95%
Boiling Point 567°C
Color White
Melting Point 193°C to 198°C
Quantity 5mg
Flash Point >110°C(230°F)
Chemical Name or Material Imipenem monohydrate
Show More Show Less

RTECSNumber : CL5446516

storageNote1 : −30° to −10°C