Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imipenem monohydrate, 98%, Acros Organics™

Click to view available options
Quantity:
250mg
1g
74431-23-5
C12H19N3O5S
317.36
GSOSVVULSKVSLQ-JJVRHELESA-N
5282372
CHEBI:51799
(5R,6S)-3-[2-(aminomethylideneamino)ethylsulfanyl]-6-[(1R)-1-hydroxyethyl]-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid;hydrate
CC(C1C2CC(=C(N2C1=O)C(=O)O)SCCN=CN)O.O
74431-23-5
C12H19N3O5S
317.36
GSOSVVULSKVSLQ-JJVRHELESA-N
5282372
CHEBI:51799
(5R,6S)-3-[2-(aminomethylideneamino)ethylsulfanyl]-6-[(1R)-1-hydroxyethyl]-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid;hydrate
CC(C1C2CC(=C(N2C1=O)C(=O)O)SCCN=CN)O.O
Infrared Spectrum Authentic
Formula Weight 317.36
Percent Purity 98%
Water 5 to 8%
Melting Point 193.0°C to 198.0°C
Quantity 1g
Chemical Name or Material Imipenem monohydrate