Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific Imipenem monohydrate, 74431-23-5, MFCD09753321, 250mg  DFS Item

Manufacturer:  Matrix Scientific 090017250MG

Molecular Formula C12H19N3O5S, Purity 95+%, Molecular Weight 317.37

Catalog No. 50-683-644


Add to cart

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit