Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imipramine Hydrochloride, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
113-52-0
C19H25ClN2
316.87
XZZXIYZZBJDEEP-UHFFFAOYSA-N
hydrogen (3-{2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-yl}propyl)dimethylamine chloride
[H+].[Cl-].CN(C)CCCN1C2=CC=CC=C2CCC2=CC=CC=C12
113-52-0
C19H25ClN2
316.87
XZZXIYZZBJDEEP-UHFFFAOYSA-N
hydrogen (3-{2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-yl}propyl)dimethylamine chloride
[H+].[Cl-].CN(C)CCCN1C2=CC=CC=C2CCC2=CC=CC=C12

IM120, 113-52-0, C19H24N2•HCl

Spectrum™ Imipramine Hydrochloride, USP is used as an antidepressant and for the treatment of nighttime bed-wetting (enuresis). All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Loss on Drying 0.005
Quantity 100 g
CAS Max % 1
Percent Purity 98 to 102%