Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Immepip dihydrobromide
Standard H3 agonist. Also H4 agonist
$197.20 - $799.24
Specifications
Chemical Name or Material | 4-(1H-Imidazol-4-ylmethyl)piperidine dihydrobromide |
---|---|
CAS | 164391-47-3 |
Solubility | Soluble to 100mM in water |
Molecular Formula | C9H15N3.2HBr |
Formula Weight | Observed MW: 327.06 |
Description
Potent histamine H3 receptor agonist. Also binds to H4 receptors (Ki values are 0.4 and 9 nM at human recombinant H3 and H4 receptors respectively). Equipotent to or slightly more active than (R)-α-methylhistamine at H3 receptors.Specifications
4-(1H-Imidazol-4-ylmethyl)piperidine dihydrobromide | |
Soluble to 100mM in water | |
Observed MW: 327.06 |
164391-47-3 | |
C9H15N3.2HBr |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title