Learn More
MilliporeSigma™ Immobilon Classico Western HRP substrate
For use on immunoblots
Manufacturer: MilliporeSigma™ WBLUC0020
Description
Western blotting, dot/slot blotting
Specifications
HRP | |
20 mL | |
2°C to 8°C, Stable for 1 year at 4°C from date of shipment. Contains hydrogen peroxide. Use in well ventilated area. |
Chemiluminescent | |
Western blotting, Dot blot, Immunoblotting (Western) | |
The reagent has been matched to optimize the range and sensitivity of detection using various antibodies in Western blot assays. |