Learn More
IMMP2L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 159701AP
Catalog No. 50-173-0609
Add to cart
Description
Description
IMP2L complex generates mature, active proteins in the mitochondrial intermembrane space by proteolytically removing the mitochondrial targeting presequence of nuclear-encoded proteins. IMP1 and IMP2 are the catalytic subunits of the IMP complex.
Specifications
Specifications
IMMP2L
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8BPT6, Q96T52
IMMP2L, IMP2, IMP2 LIKE, IMP2 like protein
Rabbit
IgG
150 μL
-20°C
Primary
100359529, 83943, 93757
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
IMMP2L
Liquid
Immp2l
IMMP2L Fusion Protein Ag8752
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat