Thermo Scientific™ Immuno Clear Standard Modules Available on GSA/VA Contract for Federal Government customers only.

Perform applications which are delivering well-to-well reproducibility with Immuno Clear Standard Modules. Manufactured to fit standard equipment.

Manufacturer: Thermo Scientific™ 6310

 View more versions of this product

Catalog No. 14-245-168

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Binding Property Adsorption,Hydrophobic
    Surface Coating Immulon 1 B,Polystyrene
    Plate Blocking None
    Format 96-well strip plate
    Detection Method Colorimetric
    Color Clear
    No. per Case 100
    No. per Pack 100
    Type Framed
    Volume (Metric) Well 380µL
    Volume (Metric) Working 330µL
    Well Design F12,F12
    Certified No
    Item Description 6310

    Perform applications which are delivering well-to-well reproducibility with Thermo Scientific™ Immuno Clear Standard Modules, which feature Nunc™, Immulon™ and Microtiter™ module plates developed for ELISA techniques. Manufactured to fit standard equipment, modules provide flexible use of components with small sample sizes. They are backed by industry-leading quality standards and certifications.

    Thermo Scientific standard modules are strip plates with solid modules that can be easily inserted and removed from their frame. This enhances the use of the plate with smaller sample sizes.

    • Several strip formats: 8-well strip, 12-well strip, 16-well strip (two columns of 8 wells)
    • Several well shapes: flat bottom, C bottom (radius edge), U shape wells, StarWells (wells have fins to increase the surface area and enhance performance)
    • Several surfaces to maximize assay performance
    • Modules remain in frame when inverted
    • Stackable frame marked for lid orientation
    • Polystyrene wells in polyethylene frame
    • Ext. Dim.: 128 x 86mm

    Choice of Surface

    • PolySorp™, Immulon 1B, and Universal Binding (UB) for adsorption of hydrophobic molecules
    • Medisorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules
    • MaxiSorp™, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays
    • MultiSorp™ are very hydrophilic and works well for glycans and water soluble proteins

    Ordering Information:

    Free-standing modules. Cat. No. 95029440 and 464394 are certified. Frames and modules available separately. Sealing tape and strip caps are available. Color-coded modules are available on request. Contact your Customer Service or Sales Representative for further details.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit