Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ImmunoCAP™ Specific IgG4 Reagents for Phadia™ 100ε

$40.00 - $154.00

Specifications

For Use With (Equipment) Phadia™ 100e
Storage Requirements 2° to 8°C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Type Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Type Price Quantity & Availability  
10-946-502P
View Documents Promotion Details
Thermo Scientific™
10946502
1 Vial x 48 Detections Specific IgG4 Conjugate
Pack of 48 for $144.00
Add to cart
 
10-946-702P
View Documents Promotion Details
Thermo Scientific™
10946702
3 Vials, 3 Runs Specific IgG4 curve control
Pack of 3 for $40.00
Add to cart
 
10-946-602P
View Documents Promotion Details
Thermo Scientific™
10946602
6 Vials, 1 Curve Specific IgG4 calibrator
Pack for $154.00
Add to cart
 
Description

Description

Specifications

Specifications

Phadia™ 100e
2° to 8°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit