Learn More
BioVision IncSupplier Diversity Partner Immunoprecipitation (IP) Kit: The Only Immunoprecipitation kit on the market that allows both IP & Co-IP of proteins/complexes as well as their functional study & analysis. Useful for tissue/cell extracts & biological samples. 25 assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K28625
Immunoprecipitation (IP) is widely used in research and development, by which a protein can be selectively purified from samples. BioVision’s immunoprecipitation kit provides optimized buffers for preparing cell/tissue extracts, antigen binding and washing steps. The protein A/G Sepharose beads provided in the kit has higher binding capacity with broader antibody isotype binding than traditional protein A or protein G resins. The kit can be used in variety of immunoprecipitation or Co-IP studies. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9667