Learn More
G-Biosciences™ Immunotag™ Human ApoB100 (Apolipoprotein B100) ELISA Kit Products for Science Education
A Sandwich ELISA Kit for the detection of Human ApoB100.
Manufacturer:  G-Biosciences™ IT1537
Catalog No. 50-148-2788
Add to cart
Description
Description
The Immunotag™ Human ApoB100 (Apolipoprotein B100) ELISA is based on a sandwich enzyme-linked immunosorbent assay (ELISA) technology. A 96-well plate is coated with a specific antibody against the protein of interest (ApoB100), which will bind the protein and this is detected by a biotin conjugated antibody against the protein of interest. The biotin moiety is subsequently detected, following washing, by the addition of streptavidin coupled horseradish peroxidase (HRP). The supplied TMB substrates were used to visualize HRP enzymatic reaction. TMB was catalyzed by HRP to produce a blue color product that changed into yellow after adding acidic stop solution. The density of yellow is directly proportional to the concentration of protein of interest captured on the plate.
Quality Assurance: Due to their limited shelf life, the ImmunoTag™ ELISA kits are not typically stocked as finished goods. Please allow 2-3 weeks for delivery. Upon receipt of an order each kit is assembled and tested to ensure that it meets specifications before shipping. Minor changes may occur to the Range, Sensitivity, and Precision and is reflected in the manual supplied with the kit (online manuals are for reference only). In the event of a significant change the order would be confirmed with the customer before shipping
Specifications
Specifications
62.5 to 4000 ng/mL
ApoB100(Apolipoprotein B100), Apo-B100
37.5 ng/mL
Sandwich ELISA, Double Antibody
Human
1 x 96-well plate
ApoB100