Learn More
G-Bioscienses Immunotag Human SFTPB (Pulmonary surfactant-associated protein B) ELISA Kit
A Sandwich ELISA Kit for the detection of Human SFTPB.
Manufacturer: G-Bioscienses IT3036
Description
The Immunotag Human SFTPB (Pulmonary surfactant-associated protein B) ELISA is based on a sandwich enzyme-linked immunosorbent assay (ELISA) technology. A 96-well plate is coated with a specific antibody against the protein of interest (SFTPB), which will bind the protein and this is detected by a biotin conjugated antibody against the protein of interest. The biotin moiety is subsequently detected, following washing, by the addition of streptavidin coupled horseradish peroxidase (HRP). The supplied TMB substrates were used to visualize HRP enzymatic reaction. TMB was catalyzed by HRP to produce a blue color product that changed into yellow after adding acidic stop solution. The density of yellow is directly proportional to the concentration of protein of interest captured on the plate.
Quality Assurance: Due to their limited shelf life, the ImmunoTag ELISA kits are not typically stocked as finished goods. Please allow 2-3 weeks for delivery. Upon receipt of an order each kit is assembled and tested to ensure that it meets specifications before shipping. Minor changes may occur to the Range, Sensitivity, and Precision and is reflected in the manual supplied with the kit (online manuals are for reference only). In the event of a significant change the order would be confirmed with the customer before shipping
Specifications
P07988 | |
0.469 ng/mL | |
SFTPB | |
1 x 96-well plate |
0.781 to 50 ng/mL | |
Human | |
SPB(Pulmonary Surfactant Associated Protein B), SFTPB, SP-B, SFTP3, Pulmonary surfactant-associated proteolipid SPL(Phe), 18 kDa pulmonary-surfactant protein, 6 kDa protein | |
Sandwich ELISA, Double Antibody |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.