Learn More
G-Bioscienses™ Immunotag™ Human WARS (Tryptophanyl tRNA Synthetase) ELISA Kit Products for Science Education
A Sandwich ELISA Kit for the detection of Human WARS.
Manufacturer:  G-Bioscienses™ IT4450
Catalog No. 50-148-5701
Add to cart
Description
Description
The Immunotag™ Human WARS (Tryptophanyl tRNA Synthetase) ELISA is based on a sandwich enzyme-linked immunosorbent assay (ELISA) technology. A 96-well plate is coated with a specific antibody against the protein of interest (WARS ), which will bind the protein and this is detected by a biotin conjugated antibody against the protein of interest. The biotin moiety is subsequently detected, following washing, by the addition of streptavidin coupled horseradish peroxidase (HRP). The supplied TMB substrates were used to visualize HRP enzymatic reaction. TMB was catalyzed by HRP to produce a blue color product that changed into yellow after adding acidic stop solution. The density of yellow is directly proportional to the concentration of protein of interest captured on the plate.
Quality Assurance: Due to their limited shelf life, the ImmunoTag™ ELISA kits are not typically stocked as finished goods. Please allow 2-3 weeks for delivery. Upon receipt of an order each kit is assembled and tested to ensure that it meets specifications before shipping. Minor changes may occur to the Range, Sensitivity, and Precision and is reflected in the manual supplied with the kit (online manuals are for reference only). In the event of a significant change the order would be confirmed with the customer before shipping
Specifications
Specifications
P23381
0.469 ng/mL
WARS
1 x 96-well plate
0.781 to 50 ng/mL
Human
WARS, FI53, WRS, TrpRS, hWRS, IFP53, Interferon-induced protein 53, Tryptophan--tRNA ligase, Tryptophanyl-tRNA synthetase, cytoplasmic
Sandwich ELISA, Double Antibody