Learn More
G-Biosciences™ Immunotag™ Mouse Hmgb1 (High mobility group protein B1) ELISA Kit

Description
The Immunotag™ Mouse Hmgb1 (High mobility group protein B1) ELISA is based on a sandwich enzyme-linked immunosorbent assay (ELISA) technology. A 96-well plate is coated with a specific antibody against the protein of interest (Hmgb1), which will bind the protein and this is detected by a biotin conjugated antibody against the protein of interest. The biotin moiety is subsequently detected, following washing, by the addition of streptavidin coupled horseradish peroxidase (HRP). The supplied TMB substrates were used to visualize HRP enzymatic reaction. TMB was catalyzed by HRP to produce a blue color product that changed into yellow after adding acidic stop solution. The density of yellow is directly proportional to the concentration of protein of interest captured on the plate.
Quality Assurance: Due to their limited shelf life, the ImmunoTag™ ELISA kits are not typically stocked as finished goods. Please allow 2-3 weeks for delivery. Upon receipt of an order each kit is assembled and tested to ensure that it meets specifications before shipping. Minor changes may occur to the Range, Sensitivity, and Precision and is reflected in the manual supplied with the kit (online manuals are for reference only). In the event of a significant change the order would be confirmed with the customer before shipping
Specifications
Specifications
| Accession Number | P63158 |
| Assay Range | 15.625 to 1000 pg/mL |
| Assay Sensitivity | 9.375 pg/mL |
| Detection Method | Sandwich ELISA, Double Antibody |
| Gene Symbol | Hmgb1 |
| Host Species | Mouse |
| Quantity | 1 x 96-well plate |
| Synonym | Hmgb1, HMG-1, Amphoterin, high mobility group box 1, High mobility group protein 1, high-mobility group(nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.