Learn More
G-Bioscienses™ Immunotag™ Rat NRG-1 (Neuregulin 1) ELISA Kit Products for Science Education
A Sandwich ELISA Kit for the detection of Rat NRG-1.
Manufacturer:  G-Bioscienses™ IT7534
Catalog No. 50-148-8767
Add to cart
Description
Description
The Immunotag™ Rat NRG-1 (Neuregulin 1) ELISA is based on a sandwich enzyme-linked immunosorbent assay (ELISA) technology. A 96-well plate is coated with a specific antibody against the protein of interest (NRG-1 ), which will bind the protein and this is detected by a biotin conjugated antibody against the protein of interest. The biotin moiety is subsequently detected, following washing, by the addition of streptavidin coupled horseradish peroxidase (HRP). The supplied TMB substrates were used to visualize HRP enzymatic reaction. TMB was catalyzed by HRP to produce a blue color product that changed into yellow after adding acidic stop solution. The density of yellow is directly proportional to the concentration of protein of interest captured on the plate.
Quality Assurance: Due to their limited shelf life, the ImmunoTag™ ELISA kits are not typically stocked as finished goods. Please allow 2-3 weeks for delivery. Upon receipt of an order each kit is assembled and tested to ensure that it meets specifications before shipping. Minor changes may occur to the Range, Sensitivity, and Precision and is reflected in the manual supplied with the kit (online manuals are for reference only). In the event of a significant change the order would be confirmed with the customer before shipping
Specifications
Specifications
Q9JKM5
0.094 ng/mL
NRG-1
1 x 96-well plate
0.156 to 10 ng/mL
Rat
NRG-1, Heregulin-1, HGL, HRG1-alpha, HRG1-beta 1, NRG1, ARIA, GGF2, GGF, glial growth factor, HGLneu differentiation factor, HRG, HRG1, HRGA, neuregulin 1 type IV beta 1a, MST131, MSTP131, NDFheregulin, alpha(45kD, ERBB2 p185-activator), neuregulin 1, neuregulin 1 type IV beta 3, pro-neuregulin-1, membrane-bound isoform, pro-NRG1, sensory and motor neuron derived factor, SMDF
Sandwich ELISA, Double Antibody