IMMY CrAg Lateral Flow Assay

Prescription-use laboratory assay aids in the diagnosis of crytococcosis

Manufacturer: IMMY MS050T

 View more versions of this product

Catalog No. 22-122-322

  Check Availability
Add to Cart

Alert

Always wear gloves when handling reagents.


Required Accessories : Pipetter (40 and 80μL); timer; disposable micro-centrifuge tubes, test tubes or microtiter plate

    Specifications

    Product Type Cryptococcus Antigen Test Reservoir Tube
    For Use With (Application) CrAg lateral flow assay
    Quantity 50 Pack
    Storage Requirements 22° to 25°C

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit