IMMY Myco DDR™ Mycobacterial Digestion/ Decontamination Kits and Components
Empowers labs to maximize the retention of viable Mycobacteria via rigid pH control throughout the digestion/decontamination process
Manufacturer: IMMY DDR0123.0
 View more versions of this product
Catalog No. 22-122-324
  Check Availability
Add to Cart
    Specifications
    Product Type 12 Specimen Reagent Set (3.0% NaOH)
    For Use With (Application) Mycobacterial testing
  • Description & Specifications
Related Products
Product Content Feedback