Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IMP2/IGF2BP2 Mouse anti-Human, Canine, Monkey, Allophycocyanin/Cy7, Clone: OTI3F9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271615APCCY7

Catalog No. N271615APC7


Add to cart

Description

Description

IMP2/IGF2BP2 Monoclonal antibody specifically detects IMP2/IGF2BP2 in Human, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

IMP2/IGF2BP2
OTI3F9
Flow Cytometry
Affinity Purified, Conjugated
IGF2BP2
Full length recombinant protein of human IMP2/IGF2BP2 (NP_006333) produced in HEK293T cells.
Immunogen affinity purified
Cancer, Chromatin Research, Immune System Diseases, Immunology, Transcription Factors and Regulators
Primary
10644.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
Hepatocellular carcinoma autoantigen p62, IGF-II mRNA-binding protein 2, IMP-2IGF2 mRNA-binding protein 2, IMP2VICKZ family member 2, insulin-like growth factor 2 mRNA binding protein 2, insulin-like growth factor 2 mRNA-binding protein 2, p62, VICKZ2
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human, Monkey
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit