Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IMPA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen IMPA2
Applications Immunofluorescence, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093350
View Documents
Novus Biologicals
NBP304631100UL
100 μg Each for $320.71
Add to cart
 
NB093351
View Documents
Novus Biologicals
NBP30463120UL
20 μg Each for $138.37
Add to cart
 
Description

Description

IMPA2 Polyclonal antibody specifically detects IMPA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

IMPA2
Unconjugated
Rabbit
Lipid and Metabolism
PBS with 50% glycerol, pH7.3.
3613
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence, Western Blot
Purified
RUO
Human, Mouse, Rat
EC 3.1.3.25, IMP 2, IMP.18P, IMPase 2, inosine monophosphatase 2, inositol monophosphatase 2, inositol monophosphatase 2 variant 1, inositol monophosphatase 2 variant 2, inositol(myo)-1(or 4)-monophosphatase 2, Inositol-1(or 4)-monophosphatase 2, Myo-inositol monophosphatase A2
Recombinant fusion protein containing a sequence corresponding to amino acids 1-288 of human IMPA2 (NP_055029.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit