Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IMPA2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP15496420UL

 View more versions of this product

Catalog No. NBP15496420


Add to cart

Description

Description

IMPA2 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

IMPA2
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
IMPA2
Synthetic peptides corresponding to IMPA2(inositol(myo)-1(or 4)-monophosphatase 2) The peptide sequence was selected from the middle region of IMPA2. Peptide sequence RGRGAFCNGQRLRVSGETDLSKALVLTEIGPKRDPATLKLFLSNMERLLH.
32 kDa
20ul
Lipid and Metabolism
Primary
3613
Western Blot
Western Blot 1:100-1:2000
O14732
EC 3.1.3.25, IMP 2, IMP.18P, IMPase 2, inosine monophosphatase 2, inositol monophosphatase 2, inositol monophosphatase 2 variant 1, inositol monophosphatase 2 variant 2, inositol(myo)-1(or 4)-monophosphatase 2, Inositol-1(or 4)-monophosphatase 2, Myo-inositol monophosphatase A2
Rabbit
IgG
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit