Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Open-Finger Shock Absorbing Glove Liners

Catalog Number 189994600b
Click to view available options
Size:
Small
Medium
Large
X-Large
Hand:
Left
Right
This item is not returnable. View return policy
This item is not returnable. View return policy

VEP padding throughout the glove liner offers protection from shock and impact. Use under work gloves in confined spaces to protect the back of the hand.

  • Breathable Nylon Lycra™ is designed to be worn inside work gloves
  • Padded with 3/16 in. impact and shock absorbing VEP pad
  • Open finger design provides tactile feedback
  • Ideal for use with hand tools, or in confined space applications such as mining
Color Black
Hand Left
Material Nylon Lycra
Product Type Glove Liners
Size Medium
Finger Style Three Quarter
For Use With (Application) Manufacturing Tools, and Confined Spaces
Glove Cuff Style Open
Series 507-01
Length (English) 9 in.
Length (Metric) 22.9 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit