Learn More
Impacto™ Open-Finger Anti-Impact Gloves with Wrist Support
VEP padding throughout the glove offers protection from shock and impact. Detachable leather wrist support prevents over flexion and over extension to help prevent repetitive strain injuries.
$44.00
Specifications
Back Material Nylon Lycra™
Cuff Material Grain Leather
Color Blue, Yellow
Material Grain Leather
Product Type Anti-Impact Gloves
View More Specs
Products 8
Catalog Number Mfr. No. Size Hand Price Quantity & Availability  
Catalog Number Mfr. No. Size Hand Price Quantity & Availability  
18-999-4610A
View Documents
Impacto™
72520110021
Small Left
Each for $44.00
Add to cart
18-999-4609A
View Documents
Impacto™
72520110022
Small Right
Each for $44.00
Add to cart
18-999-4610B
View Documents
Impacto™
72520110031
Medium Left
Each for $44.00
Add to cart
18-999-4609B
View Documents
Impacto™
72520110032
Medium Right
Each for $44.00
Add to cart
18-999-4610C
View Documents
Impacto™
72520110041
Large Left
Each for $44.00
Add to cart
18-999-4609C
View Documents
Impacto™
72520110042
Large Right
Each for $44.00
Add to cart
18-999-4610D
View Documents
Impacto™
72520110051
X-Large Left
Each for $44.00
Add to cart
18-999-4609D
View Documents
Impacto™
72520120052
X-Large Right
Each for $44.00
Add to cart
Description
Description
  • Contoured VEP 1/8 in. padding in the palm extends to the wrist joint, fingers, and thumb web area to protect from impact and shock
  • Detachable leather wrist support prevents over flexion and over extension to help prevent RSI
  • Nylon fabric on the back ensures optimum breathability and comfort
  • Open-finger design allows for tactile feedback
  • For protection from hand tools and power tools in applications including construction, assembly, manufacturing, and maintenance
Specifications
Specifications
Nylon Lycra™
Blue, Yellow
Anti-Impact Gloves
Construction, Assembly, Manufacturing, and Maintenance
725-20
Grain Leather
Grain Leather
Three Quarter
Yellow