Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Impacto™ Full-Finger Anti-Impact Grain Leather Gloves
Click to view available options
Size:
Small
Medium
Large
X-Large
Hand:
Left
Right
Description
- Durable grain leather offers abrasion protection and ensures durability
- Contoured VEP 1/8 in. padding in the palm, fingers, and thumb protects from impact
- Full finger style for protection of the entire hand
- 100% aramid fiber stitching increases the glove's durability
- Elastic gathering at the wrist helps to prevent debris from getting inside of the glove
- Safety suede leather cuff with outer seams to ensure user comfort
- Protection from air and power tools in applications such as construction, and maintenance
Specifications
Cuff Material | Suede Leather |
Color | Yellow |
Hand | Right |
Material | Grain Leather |
Product Type | Anti-Impact Gloves |
Size | Small |
Finger Style | Full |
For Use With (Application) | Construction, Maintenance |
Glove Cuff Color | White |
Series | 615-20 |
Show More |