Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Full-Finger Anti-Impact Grain Leather Gloves

Click to view available options
Size:
Small
Medium
Large
X-Large
Hand:
Left
Right
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves are stitched with DuPont™ Aramid Fibers for durability and abrasion protection. VEP padding in the palm, fingers, and thumb provides users with impact protection.

  • Durable grain leather offers abrasion protection and ensures durability
  • Contoured VEP 1/8 in. padding in the palm, fingers, and thumb protects from impact
  • Full finger style for protection of the entire hand
  • 100% aramid fiber stitching increases the glove's durability
  • Elastic gathering at the wrist helps to prevent debris from getting inside of the glove
  • Safety suede leather cuff with outer seams to ensure user comfort
  • Protection from air and power tools in applications such as construction, and maintenance
Cuff Material Suede Leather
Color Yellow
Hand Right
Material Grain Leather
Product Type Anti-Impact Gloves
Size X-Large
Finger Style Full
For Use With (Application) Construction, Maintenance
Glove Cuff Color White
Series 615-20
Length (English) 11 in.
Length (Metric) 28 cm
Show More Show Less