Impacto™ Anti-Impact Gloves 400 Series

Manufacturer: Impacto Protective Products AV40930RL

 View more versions of this product

Catalog No. 18-999-4595C

  Check Availability
Add to Cart

    Specifications

    Certifications EN420 and EN388
    Circumference (English) Hand 9 to 10 in.
    Glove Cuff Style Gauntlet
    For Use With (Application) For work in warm environments or as a liner under specialty work gloves
    Material Nylon Lycra™/Leather/Polycotton
    Size 9 (Large)
    Color Black and White

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit