Learn More
Impacto™ Anti-Impact Trigger Finger Glove
Protection from vibration and impact
Manufacturer:  Impacto™ 40930 LG RH
 View more versions of this product
Catalog No. 19-048-713F
Add to cart
Description
Description
  • Half finger ensures tactile feedback; index finger is extended to protect from tool impact and vibration
  • Nylon Lycra back for comfort, dexterity and moisture evaporation
  • Soft pearl leather palm and fingers
  • Impact absorbing visco-elastic polymer padding (VEP) in palm, trigger finger and thumb
  • Adjustable elastic wrist with hook and loop closure
  • Fingertips have pull tabs for easy removal
Specifications
Specifications
Safety
23 to 25cm
Half
Right
Leather, Lycra™
9 in.
9 to 10 in.
Nylon Lycra™, Pearl Leather
Soft
Visco Elastic Polymer padding in palm, thumb, trigger finger, Elastic wrist closure
Large
23 cm