Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Double Padded Anti-Impact Gloves

Catalog Number 189994605A
Click to view available options
Size:
Small
Medium
Large
X-Large
Hand:
Left
Right
This item is not returnable. View return policy
This item is not returnable. View return policy

Nylon material offers breathability and comfort, while suede leather provides abrasion protection and durability. 1/4 in. VEP padding in the palm extends to the edge of the hand for protection from impact and shock.

  • VEP provides impact absorbing and shock dissipation qualities, as well as absorption of higher levels of vibration
  • Fingerless style for tactile feedback and mobility
  • Suede leather offers abrasion protection & durability
  • Side padding protects from impact during automotive applications
Color Blue, Yellow
Hand Right
Material Nylon Lycra
Product Type Anti-impact Gloves
Size Small
Finger Style Fingerless
For Use With (Application) Automobile Construction
Glove Cuff Style Open
Series 523-24
Length (English) 7 in.
Length (Metric) 18 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit