Learn More
Impacto™ Protective Elbow Pad
Impacto™ 806-20 Elbow protector padded protect from cumulative trauma such as bruising
Supplier: Impacto™ 80610110030
Description
Impacto™ 806-10 Elbow Protector
- Stretchy polyester with durable suede leather cover for abrasion protection
- Layered impact absorbing Visco-elastic polymer padding
- Helps prevent cumulative trauma
- Provides impact, shock and abrasion protection
- Offers cushioning and protection from bruising
- Available with suede leather cover and grain leather cover
Order Info
Split and full grain leather versions available.
Specifications
Pull-on | |
Medium | |
Suede |
Elbow Pad | |
Offers cushioning and protection from bruising |