Impacto Fingerless Glove Liners with Grain Leather Wrist Support

Manufacturer: Impacto Protective Products AV71420PLRG

 View more versions of this product

Catalog No. 19-039268

  Check Availability
Add to Cart

    Specifications

    Color Blue. gold
    Hand Ambidextrous
    Includes Detachable grain Leather wrist support, VEP palm padding
    Material Polycotton Fabric
    Closure Type Hook and loop
    For Use With (Application) Provides protection from impact and shock and recommended to be worn inside specialty glove for maximum durability
    Style Fingerless
    Product Type Fingerless Glove Liner
    Size Large

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit